Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531266
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (87%), Dog (91%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Deubiquitinase that antagonizes the anaphase-promoting complex (APC/C) during G1/S transition by mediating deubiquitination of cyclin-A (CCNA1 and CCNA2), thereby promoting S phase entry. Specifically mediates deubiquitination of 'Lys-11'-linked polyubiquitin chains, a specific ubiquitin-linkage type mediated by the APC/C complex. Also mediates deubiquitination of 'Lys-48'-linked polyubiquitin chains in vitro. Phosphorylation at Ser-628 during G1/S phase maximizes the deubiquitinase activity, leading to prevent degradation of cyclin-A (CCNA1 and CCNA2) (PubMed:21596315). Plays an important role in the regulation of DNA replication by stabilizing the licensing factor CDT1 (PubMed:27296872). [UniProt].
Specifications
| USP37 | |
| Polyclonal | |
| Unconjugated | |
| Usp37 | |
| 4932415L06Rik; C330008N13Rik; Deubiquitinating enzyme 37; Kiaa1594; tmp_locus_50; ubiquitin carboxyl-terminal hydrolase 37; ubiquitin specific peptidase 37; ubiquitin specific peptidase 37-like; ubiquitin specific protease 37; ubiquitin thioesterase 37; ubiquitin thiolesterase 37; ubiquitin-specific-processing protease 37; USP37 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57695 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q86T82 | |
| Usp37 | |
| Recombinant protein encompassing a sequence within the center region of human USP37. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction