Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP37 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | USP37 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2156
![]() ![]() |
Proteintech Group Inc
184651AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The ubiquitin (Ub) pathway involves three sequential enzymatic steps that facilitate the conjugation of Ub and Ub-like molecules to specific protein substrates. Through the use of a wide range of enzymes that can add or remove ubiquitin, the Ub pathway controls many intracellular processes such as signal transduction, transcriptional activation and cell cycle progression. USP37 (ubiquitin specific peptidase 37) is a 979 amino acid protein that contains three UIM (ubiquitin-interacting motif) repeats and belongs to thepeptidase C19 family. Expressed in prostate and brain tissue, USP37 functions to catalyze the conversion of a ubiquitin C-terminal thioester to a free ubiquitin and a thiol. The gene encoding USP37 maps to human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome.Specifications
USP37 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q86T82, Q8C0R0 | |
100361658, 319651, 57695 | |
USP37 Fusion Protein Ag12513 | |
Primary | |
-20°C | |
USP37 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 37, KIAA1594, Ubiquitin thiolesterase 37, USP37 | |
Usp37 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title