Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP38 Monoclonal Antibody (OTI1D11)
GREENER_CHOICE

Catalog No. PIMA524903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524903 Supplier Invitrogen™ Supplier No. MA524903
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP38 Monoclonal Antibody for Western Blot

Deubiquitinating enzyme exhibiting a preference towards 'Lys-63'-linked Ubiquitin chains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP38
Applications Western Blot
Classification Monoclonal
Clone OTI1D11
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Usp38
Gene Accession No. Q8NB14
Gene Alias 4631402N15Rik; 4833420O05Rik; AA536967; AU044701; AW544820; deubiquitinating enzyme 38; HP43.8KD; KIAA1891; mKIAA1891; ubiquitin carboxyl-terminal hydrolase 38; ubiquitin specific peptidase 38; ubiquitin specific protease 38; ubiquitin thioesterase 38; ubiquitin thiolesterase 38; ubiquitin-specific-processing protease 38; Usp38
Gene Symbols Usp38
Host Species Mouse
Immunogen Full length human recombinant protein of USP38 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 476071, 84640
Target Species Human, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.