Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565184
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565184 Supplier Invitrogen™ Supplier No. PA565184
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEHWLDEAQCE AMFDLTTRLI LEGQDPFQRQ VGHQVLEAYA RYHRPEFESF FNKTFVLGLL HQGYHSLDRK DVAILDYIHN GLKLIMSCPS Highest antigen sequence identity to the following orthologs - mouse 98%, rat 99%.

Deubiquitinating enzyme exhibiting a preference towards 'Lys-63'-linked Ubiquitin chains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP38
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Usp38
Gene Accession No. Q8NB14
Gene Alias 4631402N15Rik; 4833420O05Rik; AA536967; AU044701; AW544820; deubiquitinating enzyme 38; HP43.8KD; KIAA1891; mKIAA1891; ubiquitin carboxyl-terminal hydrolase 38; ubiquitin specific peptidase 38; ubiquitin specific protease 38; ubiquitin thioesterase 38; ubiquitin thiolesterase 38; ubiquitin-specific-processing protease 38; Usp38
Gene Symbols Usp38
Host Species Rabbit
Immunogen Recombinant Human USP38. Recombinant protein control fragment (Product #RP-105931).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84640
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.