Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP39 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA597229

Catalog No. PIPA597229


Only null left
Add to Cart

Description

Description

Immunogen sequence: GTKKKKKTIV TDVFQGSMRI FTKKLPHPDL PAEEKEQLLH NDEYQETMVE STFMYLTLDL PTAPLYKDEK EQLIIPQVPL FNILAKFNGI TEKEYKTYKE NFLKRFQLTK LPPYLIFCIK RFTKNNFFVE KNPTIVNFPI TNVDLREYLS EEVQAVHKNT TYDLIANIVH DGKPSEGSYR IHVLHHGTGK WYELQDLQVT DILPQMITLS EAYIQIWKRR DNDETNQQGA; Positive Samples: HepG2, HeLa, A-375, HT-29, B-cell, Mouse eye, Mouse spleen, Rat testis; Cellular Location: Nucleus.

USP39 may play a role in mRNA splicing. It is unsure if the protein really exhibits hydrolase activity. It could be a competitor of ubiquitin C-terminal hydrolases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP39
Polyclonal
Unconjugated
USP39
65K; AA408960; AI894154; CGI-21; D6Wsu157e; HSPC332; Inactive ubiquitin-specific peptidase 39; PRO2855; SAD1; SAD1 homolog; small nuclear ribonucleoprotein 65kDa (U4/U6.U5); SnRNP assembly defective 1 homolog; SNRNP65; U4/U6.U5 tri-snRNP-associated 65 kDa protein; U4/U6.U5 tri-snRNP-associated protein 2; ubiquitin specific peptidase 39; ubiquitin specific protease 39; Usp39
Rabbit
Affinity Chromatography
RUO
10713, 28035, 297336
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.86 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q3TIX9, Q53GS9
USP39
Recombinant fusion protein containing a sequence corresponding to amino acids 336-565 of human USP39 (NP_0065812).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.