Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP39 Recombinant Rabbit Monoclonal Antibody (9E7O2)
GREENER_CHOICE

Catalog No. PIMA542666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542666 Supplier Invitrogen™ Supplier No. MA542666
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HCT116. The target is usually found in the following locations: Nucleus. Immunogen sequence: QQIANLDKQA KLSRAYDGTT YLPGIVGLNN IKANDYANAV LQALSNVPPL RNYFLEEDNY KNIKRPPGDI MFLLVQRFGE LMRKLWNPRN FKAHVSPHEM L.

USP39 may play a role in mRNA splicing. It is unsure if the protein really exhibits hydrolase activity. It could be a competitor of ubiquitin C-terminal hydrolases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP39
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 9E7O2
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene USP39
Gene Accession No. Q53GS9
Gene Alias 65K; AA408960; AI894154; CGI-21; D6Wsu157e; HSPC332; Inactive ubiquitin-specific peptidase 39; PRO2855; SAD1; SAD1 homolog; small nuclear ribonucleoprotein 65kDa (U4/U6.U5); SnRNP assembly defective 1 homolog; SNRNP65; U4/U6.U5 tri-snRNP-associated 65 kDa protein; U4/U6.U5 tri-snRNP-associated protein 2; ubiquitin specific peptidase 39; ubiquitin specific protease 39; Usp39
Gene Symbols USP39
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human USP39 (Q53GS9).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10713, 297336
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.