Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP4 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002869JF646
Description
USP4 Polyclonal antibody specifically detects USP4 in Human samples. It is validated for Western BlotSpecifications
USP4 | |
Polyclonal | |
Western Blot | |
Deubiquitinating enzyme 4, EC 3.1.2.15, EC 3.4.19.12, MGC149848, MGC149849, ubiquitin carboxyl-terminal esterase 4, ubiquitin carboxyl-terminal hydrolase 4, ubiquitin specific peptidase 4 (proto-oncogene), ubiquitin specific protease 4 (proto-oncogene), ubiquitin thioesterase 4, Ubiquitin thiolesterase 4, ubiquitin-specific processing protease 4, Ubiquitin-specific-processing protease 4, Ubiquitous nuclear protein homolog, UNPH, UNPUnph | |
The immunogen recognized by this antibody maps to a region between residue 925 and the C-terminus (residue 963) of human Ubiquitin Specific Protease 4 using the numbering given in entry AAB72237.1 (GeneID 7375). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7375 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction