Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533106
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533106 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533106 Supplier Invitrogen™ Supplier No. PA533106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Dog, Bat, Bovine, Elephant, Panda, Horse, Rabbit, Pig (100%); Hamster, Opossum (94%); Mouse, Rat, Turkey, Platypus (89%); Chicken, Xenopus (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

The protein encoded by this gene is a protease that deubiquitinates target proteins such as ADORA2A and TRIM21. The encoded protein shuttles between the nucleus and cytoplasm and is involved in maintaining operational fidelity in the endoplasmic reticulum. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene USP4
Gene Accession No. A6QR55, Q13107
Gene Alias Deubiquitinating enzyme 4; F730026I20Rik; mKIAA4155; ubiquitin carboxyl-terminal esterase 4; ubiquitin carboxyl-terminal hydrolase 4; ubiquitin specific peptidase 4; ubiquitin specific peptidase 4 (proto-oncogene); ubiquitin specific protease 4 (proto-oncogene); ubiquitin thioesterase 4; ubiquitin thiolesterase 4; ubiquitin-specific processing protease 4; ubiquitin-specific-processing protease 4; Ubiquitous nuclear protein; ubiquitous nuclear protein homolog; Unp; Unph; USP4
Gene Symbols USP4
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human USP4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100053443, 100359136, 100512072, 476624, 508042, 7375
Target Species Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.