Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599736 Supplier Invitrogen™ Supplier No. PA599736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total USP40.

May be catalytically inactive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP40
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP40
Gene Accession No. Q8BWR4, Q9NVE5
Gene Alias B230215L03Rik; C730029K03; deubiquitinating enzyme 40; LRRGT00071; Ubiquitin carboxyl-terminal hydrolase 40; ubiquitin specific peptidase 40; ubiquitin specific protease 40; ubiquitin thioesterase 40; ubiquitin thiolesterase 40; ubiquitin-specific-processing protease 40; USP40
Gene Symbols USP40
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP40(Accession Q9NVE5), corresponding to amino acid residues F1013-N1063.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227334, 316599, 55230
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.