Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP40 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548596
Description
Predicted to react with mouse based on sequence homology.
May be catalytically inactive.
Specifications
USP40 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8BWR4 | |
USP40 | |
KLH conjugated synthetic peptide between 63-91 amino acids from the N-terminal region of human USP40. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Usp40 | |
Ubiquitin carboxyl-terminal hydrolase 40, Deubiquitinating enzyme 40, Ubiquitin thioesterase 40, Ubiquitin-specific-processing protease 40, USP40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
227334 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction