Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536513
Description
This antibody detects endogenous protein at a molecular weight of 150 kDa. Purity is >95% by SDS-PAGE.
USP42 (ubiquitin specific peptidase 42) is a 1,325 amino acid protein that belongs to the peptidase C19 family. Expressed in a variety of tissues, USP42 functions to catalyze the conversion of a ubiquitin C-terminal thioester to a free ubiquitin and a thiol. The gene encoding USP42 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
Specifications
USP42 | |
Polyclonal | |
Unconjugated | |
Usp42 | |
2410140K03Rik; 3110031A07Rik; A630018G05Rik; D5Ertd591e; Deubiquitinating enzyme 42; ubiquitin carboxyl-terminal hydrolase 42; ubiquitin specific peptidase 42; ubiquitin specific protease 42; ubiquitin thioesterase 42; ubiquitin thiolesterase 42; ubiquitin-specific-processing protease 42; Usp42 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288482, 76800, 84132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
B2RQC2, D3ZU96, Q9H9J4 | |
Usp42 | |
Synthetic peptide corresponding to amino acids 254-306 of human USP42. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction