Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101945
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101945 Supplier Invitrogen™ Supplier No. PA5101945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total USP43.

May recognize and hydrolyze the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP43
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP43
Gene Accession No. Q70EL4, Q8BUM9
Gene Alias Deubiquitinating enzyme 43; ubiquitin carboxyl-terminal hydrolase 43; ubiquitin specific peptidase 43; ubiquitin specific protease 43; ubiquitin thioesterase 43; ubiquitin thiolesterase 43; ubiquitin-specific-processing protease 43; USP43
Gene Symbols USP43
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP43(Accession Q70EL4), corresponding to amino acid residues N113-L163.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124739, 216835
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.