Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144738 Supplier Invitrogen™ Supplier No. PA5144738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEDLNTIAEG DNVYAFQVPP SPSQGTLSAH PLGLSASPRL AAREGQRFSL SLHSESKVLI LFCNLVGSGQ QASRFGPPFL IREDRAVSWA QLQQSILSKV RHLMKSEAPV QNLGSLFSIR VVGLSVACSY LSPKD Antigen sequence identity: Mouse 81%; Rat 76%.

May recognize and hydrolyze the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP43
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP43
Gene Accession No. Q70EL4
Gene Alias Deubiquitinating enzyme 43; ubiquitin carboxyl-terminal hydrolase 43; ubiquitin specific peptidase 43; ubiquitin specific protease 43; ubiquitin thioesterase 43; ubiquitin thiolesterase 43; ubiquitin-specific-processing protease 43; USP43
Gene Symbols USP43
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 341-475. Recombinant protein control fragment (Product #RP-110170).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124739
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.