Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP43 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548680
Description
May recognize and hydrolyze the peptide bond at the C- terminal Gly of ubiquitin. Involved in the processing of poly- ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications
USP43 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q70EL4 | |
USP43 | |
KLH conjugated synthetic peptide between 900-928 amino acids from the C-terminal region of human USP43. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
USP43 | |
Ubiquitin carboxyl-terminal hydrolase 43, Deubiquitinating enzyme 43, Ubiquitin thioesterase 43, Ubiquitin-specific-processing protease 43, USP43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124739 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction