Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP44 Monoclonal Antibody (OTI1F9)
GREENER_CHOICE

Catalog No. PIMA526519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526519 Supplier Invitrogen™ Supplier No. MA526519
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP44 Monoclonal Antibody for Western Blot, IHC (P)

Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP44 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP44
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene USP44
Gene Accession No. Q9H0E7
Gene Alias Deubiquitinating enzyme 44; E430004F17Rik; hypothetical protein MGC55661; inactive deubiquitinating enzyme 44; inactive ubiquitin carboxyl-terminal hydrolase 44; inactive ubiquitin thioesterase 44; inactive ubiquitin-specific-processing protease 44; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 44; ubiquitin carboxyl-terminal hydrolase 44; ubiquitin specific peptidase 44; ubiquitin specific peptidase 49 pseudogene; ubiquitin specific protease 44; ubiquitin thioesterase 44; ubiquitin thiolesterase 44; ubiquitin-specific-processing protease 44; USP44; zgc:55661
Gene Symbols USP44
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 420-712 of USP44 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84101
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.