Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556205 Supplier Invitrogen™ Supplier No. PA556205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKAKRSKRPT VPHDEDSSDD IAVGLTCQHV SHAISVNHVK RAIAENLWSV CSECLEERRF YDGQLVLTSD IWLCLKCGFQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.

The protein encoded by this gene is a deubiquitylase that binds ERCC1, the catalytic subunit of the XPF-ERCC1 DNA repair endonuclease. This endonuclease is a critical regulator of DNA repair processes, and the deubiquitylase activity of the encoded protein is important for maintaining the DNA repair ability of XPF-ERCC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP45
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Usp45
Gene Accession No. Q70EL2
Gene Alias 3110003C05Rik; 4930550B20Rik; AI843191; Deubiquitinating enzyme 45; Gcap7; granule cell antiserum positive 7; ubiquitin carboxyl-terminal hydrolase 45; ubiquitin specific peptidase 45; ubiquitin specific petidase 45; ubiquitin specific protease 45; ubiquitin thioesterase 45; ubiquitin thiolesterase 45; ubiquitin-specific-processing protease 45; USP45
Gene Symbols Usp45
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP45. Recombinant protein control fragment (Product #RP-94741).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.