Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP46 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP182293
Description
USP46 Polyclonal specifically detects USP46 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
USP46 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200 - 1:500, Chromatin Immunoprecipitation (ChIP) | |
P62068 | |
USP46 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LFDNYMQQDAHEFLNYLLNTIADILQEEKKQEKQNGKLKNGNMNEPAENNKPELTWVHEIFQGTLTNETRCLNCETVSSKDEDFLDLSVDVEQN | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP Assay | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Deubiquitinating enzyme 46, EC 3.1.2.15, EC 3.4.19.12, FLJ11850, FLJ12552, FLJ14283, FLJ39393, ubiquitin carboxyl-terminal hydrolase 46, ubiquitin specific peptidase 46, ubiquitin specific protease 46, ubiquitin thioesterase 46, Ubiquitin thiolesterase 46, Ubiquitin-specific-processing protease 46 | |
Rabbit | |
42 kDa | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
64854 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction