Learn More
Invitrogen™ USP46 Monoclonal Antibody (CL0363)

Description
Immunogen sequence: LFDNYMQQDA HEFLNYLLNT IADILQEEKK QEKQNGKLKN GNMNEPAENN KPELTWVHEI FQGTLTNETR CLNCETVSSK DEDFLDLSVD VEQN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%. Binds to an epitope located within the peptide sequence TNETRCLNCE as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | USP46 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL0363 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | USP46 |
| Gene Accession No. | P62068 |
| Gene Alias | 1190009E20Rik; 2410018I08Rik; AI451644; Deubiquitinating enzyme 46; RGD1564808; Ubiquitin carboxyl-terminal hydrolase 46; ubiquitin specific peptidase 46; ubiquitin specific protease 46; ubiquitin thioesterase 46; ubiquitin thiolesterase 46; ubiquitin-specific-processing protease 46; Usp46 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.