Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581110 Supplier Invitrogen™ Supplier No. PA581110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human USP46.

Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP46
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene USP46
Gene Accession No. P62068, P62069
Gene Alias 1190009E20Rik; 2410018I08Rik; AI451644; Deubiquitinating enzyme 46; RGD1564808; Ubiquitin carboxyl-terminal hydrolase 46; ubiquitin specific peptidase 46; ubiquitin specific protease 46; ubiquitin thioesterase 46; ubiquitin thiolesterase 46; ubiquitin-specific-processing protease 46; Usp46
Gene Symbols USP46
Host Species Rabbit
Immunogen Recombinant Human USP46 Protein (Met1-Glu366).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64854, 69727
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.