Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104201 Supplier Invitrogen™ Supplier No. PA5104201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total USP46.

Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP46
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP46
Gene Accession No. F1M625, P62068, P62069
Gene Alias 1190009E20Rik; 2410018I08Rik; AI451644; Deubiquitinating enzyme 46; RGD1564808; Ubiquitin carboxyl-terminal hydrolase 46; ubiquitin specific peptidase 46; ubiquitin specific protease 46; ubiquitin thioesterase 46; ubiquitin thiolesterase 46; ubiquitin-specific-processing protease 46; Usp46
Gene Symbols USP46
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP46(Accession P62068), corresponding to amino acid residues N126-L176.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289584, 64854, 69727
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.