Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110560
Description
Immunogen sequence: KKFISRLRKE NDLFDNYMQQ DAHEFLNYLL NTIADILQEE KKQEKQNGKL KNGNMNEPAE NNKPELTWVH EIFQGTLTNE TRCLNCETVS SKDEDFLDLS V.
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
Specifications
USP46 | |
Polyclonal | |
Unconjugated | |
USP46 | |
1190009E20Rik; 2410018I08Rik; AI451644; Deubiquitinating enzyme 46; RGD1564808; Ubiquitin carboxyl-terminal hydrolase 46; ubiquitin specific peptidase 46; ubiquitin specific protease 46; ubiquitin thioesterase 46; ubiquitin thiolesterase 46; ubiquitin-specific-processing protease 46; Usp46 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
289584, 64854, 69727 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
F1M625, P62068, P62069 | |
USP46 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human USP46 (NP_0737432). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction