Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP47 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057487F
Description
USP47 Polyclonal antibody specifically detects USP47 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
USP47 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Deubiquitinating enzyme 47, DKFZp686C13257, EC 3.4.19.12, FLJ20727, Trf (TATA binding protein-related factor)-proximal homolog, Trf (TATA binding protein-related factor)-proximal homolog (Drosophila), TRFP, ubiquitin carboxyl-terminal hydrolase 47, ubiquitin specific peptidase 47, ubiquitin specific protease 47, ubiquitin thioesterase 47, Ubiquitin thiolesterase 47, Ubiquitin-specific-processing protease 47 | |
The immunogen recognized by this antibody maps to a region between residue 1237 and the C-terminus (residue 1287) of human ubiquitin specific peptidase 47 using the numbering given in entry NP_060414.3 (GeneID 55031). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55031 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction