Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP47 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559113
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9113 Supplier Bethyl Laboratories, Inc Supplier No. A301049A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Chicken

Putative ubiquitin-specific-processing protease that regulates cell growth and survival. Probably regulates CDC25A expression at a transcriptional level. May be catalytically inactive although it seems to have kept all necessary active site residues.

Specifications

Antigen USP47
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene USP47
Gene Accession No. Q96K76
Gene Alias TRFP, Trf (TATA binding protein-related factor)-proximal homolog, deubiquitinating enzyme 47, ubiquitin carboxyl-terminal hydrolase 47, ubiquitin specific protease 47, ubiquitin thioesterase 47, ubiquitin thiolesterase 47, ubiquitin-specific-processing protease 47
Gene Symbols USP47
Host Species Rabbit
Immunogen Between 1237 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55031
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.