Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP48 Monoclonal Antibody (OTI1F10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF801869


Description
USP48 Monoclonal antibody specifically detects USP48 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
USP48 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P56703 | |
Mouse | |
Protein A/G | |
RUO | |
7473 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1F10 | |
Unconjugated | |
WNT3 | |
2810449C13Rik; AI115503; BC021769; D330022K21Rik; Deubiquitinating enzyme 48; Kiaa4202; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 48; RAP1GA1; Synaptic ubiquitin-specific protease; synUSP; Ubiquitin carboxyl-terminal hydrolase 48; Ubiquitin carboxyl-terminal hydrolase 48-like protein; ubiquitin specific peptidase 48; ubiquitin specific protease 31; ubiquitin specific protease 48; ubiquitin thioesterase 48; ubiquitin thiolesterase 48; Ubiquitin-specific peptidase 48; Ubiquitin-specific protease 48; Ubiquitin-specific-processing protease 48; Usp31; Usp48 | |
Human recombinant protein fragment corresponding to amino acids 128-355 of human WNT3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction