Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP48 Mouse anti-Human, Clone: OTI1F10, TrueMAB™, Origene
SDP

Catalog No. 502243291
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-224-3291 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-224-3291 Supplier OriGene Supplier No. TA801869S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP48 Monoclonal antibody specifically detects USP48 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen USP48
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F10
Concentration 1.022 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene USP48
Gene Accession No. Q86UV5
Gene Alias 2810449C13Rik; AI115503; BC021769; D330022K21Rik; Deubiquitinating enzyme 48; Kiaa4202; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 48; RAP1GA1; Synaptic ubiquitin-specific protease; synUSP; Ubiquitin carboxyl-terminal hydrolase 48; Ubiquitin carboxyl-terminal hydrolase 48-like protein; ubiquitin specific peptidase 48; ubiquitin specific protease 31; ubiquitin specific protease 48; ubiquitin thioesterase 48; ubiquitin thiolesterase 48; Ubiquitin-specific peptidase 48; Ubiquitin-specific protease 48; Ubiquitin-specific-processing protease 48; Usp31; Usp48
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 361-743 of human USP48 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84196
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.