Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120897 Supplier Invitrogen™ Supplier No. PA5120897
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, HT-29, HeLa, A375, Mouse lung, Rat brain. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MAPRLQLEKA AWRWAETVRP EEVSQEHIET AYRIWLEPCI RGVCRRNCKG NPNCLVGIGE HIWLGEIDEN SFHNIDDPNC ERRKKNSFVG LTNLGATCYV NTFLQVWFLN LELRQALYLC PSTCSDYMLG DGIQEEKDYE PQTICEHLQY LFALLQNSNR RYIDPSGFVK ALGLDTGQQQ DAQEFSKLFM SLLEDTLSKQ KNPDVRNIVQ QQFCGEYAYV TVCNQCGRES KLLSKFYELE LNIQGHKQLT DCISEFLKEE KLEGDNRYFC ENCQSKQNAT RKIRLLSLPC TLNLQLMRFV.

USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP48
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene USP48
Gene Accession No. Q3V0C5, Q76LT8, Q86UV5
Gene Alias 2810449C13Rik; AI115503; BC021769; D330022K21Rik; Deubiquitinating enzyme 48; Kiaa4202; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 48; RAP1GA1; Synaptic ubiquitin-specific protease; synUSP; Ubiquitin carboxyl-terminal hydrolase 48; Ubiquitin carboxyl-terminal hydrolase 48-like protein; ubiquitin specific peptidase 48; ubiquitin specific protease 31; ubiquitin specific protease 48; ubiquitin thioesterase 48; ubiquitin thiolesterase 48; Ubiquitin-specific peptidase 48; Ubiquitin-specific protease 48; Ubiquitin-specific-processing protease 48; Usp31; Usp48
Gene Symbols USP48
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human USP48 (NP_115612.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170707, 362636, 84196
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.