Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576537
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications
USP48 | |
Polyclonal | |
Unconjugated | |
USP48 | |
2810449C13Rik; AI115503; BC021769; D330022K21Rik; Deubiquitinating enzyme 48; Kiaa4202; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 48; RAP1GA1; Synaptic ubiquitin-specific protease; synUSP; Ubiquitin carboxyl-terminal hydrolase 48; Ubiquitin carboxyl-terminal hydrolase 48-like protein; ubiquitin specific peptidase 48; ubiquitin specific protease 31; ubiquitin specific protease 48; ubiquitin thioesterase 48; ubiquitin thiolesterase 48; Ubiquitin-specific peptidase 48; Ubiquitin-specific protease 48; Ubiquitin-specific-processing protease 48; Usp31; Usp48 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170707, 362636, 84196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3V0C5, Q76LT8, Q86UV5 | |
USP48 | |
Recombinant fusion protein of human USP48. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction