Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568357 Supplier Invitrogen™ Supplier No. PA568357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Specifically deubiquitinates histone H2B at 'Lys-120' (H2BK120Ub). H2BK120Ub is a specific tag for epigenetic transcriptional activation and acts as a regulator of mRNA splicing. Deubiquitination is required for efficient cotranscriptional splicing of a large set of exons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP49
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP49
Gene Accession No. Q6P9L4, Q70CQ1
Gene Alias C330046L10Rik; deubiquitinating enzyme 49; Gm545; RGD1310513; ubiquitin carboxyl-terminal hydrolase 49; ubiquitin specific peptidase 49; ubiquitin specific protease 49; ubiquitin thioesterase 49; ubiquitin thiolesterase 49; ubiquitin-specific-processing protease 49; Usp49
Gene Symbols USP49
Host Species Rabbit
Immunogen Synthesized peptide derived from human USP49.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224836, 25862, 316211
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.