Learn More
USP5/IsoT Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
| Antigen | USP5/IsoT |
|---|---|
| Applications | Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1882
|
Bethyl Laboratories, Inc
IHC00313T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1881
|
Bethyl Laboratories, Inc
IHC00313 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Crab-eating macaque and Wh
USP5 (ubiquitin carboxyl-terminal hydrolase 5) cleaves linear and branched multiubiquitin polymers with a marked preference for branched polymers. It is involved in uchored Lys-48-linked polyubiquitin disassembly, and also binds linear and Lys-63 linked polyubiquitin with a lower affinity. Knock-down of USP5 causes the accumulation of p53/TP53 and an increase in p53/TP53 transcriptional activity because the uchored polyubiquitin that accumulates is able to compete with ubiquitinated p53/TP53 but not with MDM2 for proteasomal recognition.Specifications
| USP5/IsoT | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Ubiquitin carboxyl-terminal hydrolase 5; Ubiquitin thiolesterase 5; Ubiquitin-specific processing protease 5; Deubiquitinating enzyme 5; Isopeptidase T | |
| USP5 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P45974 | |
| 8078 | |
| Between 375 and 425 | |
| Primary | |
| 4° C | |
| USP5 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.