Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP5 Monoclonal Antibody (OTI2E3), TrueMAB™, OriGene
SDP

Catalog No. 502542405
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2405 30 μL
50-167-2438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2405 Supplier OriGene Supplier No. TA501290S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP5 Monoclonal antibody specifically detects USP5 in Canine, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen USP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2E3
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene USP5
Gene Accession No. P45974
Gene Alias AA407472; deubiquitinating enzyme 5; EGK_03293; Isopeptidase T; Isot; ISOT-1; testicular tissue protein Li 218; ubiquitin carboxy terminal hydrolase T; ubiquitin carboxyl-terminal hydrolase 5; ubiquitin isopeptidase T; ubiquitin specific peptidase 5; ubiquitin specific peptidase 5 (isopeptidase T); ubiquitin specific protease 5; ubiquitin specific protease 5 (isopeptidase T); ubiquitin thioesterase 5; ubiquitin thiolesterase 5; ubiquitin-specific protease-5 (ubiquitin isopeptidase T); ubiquitin-specific-processing protease 5; Ucht; Usp5
Host Species Mouse
Immunogen Full length human recombinant protein of human USP5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297593, 486718, 8078
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.