Learn More
USP5 Mouse anti-Canine, Human, Rat, Clone: OTI2E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501290

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
USP5 (ubiquitin carboxyl-terminal hydrolase 5) cleaves linear and branched multiubiquitin polymers with a marked preference for branched polymers. It is involved in uchored Lys-48-linked polyubiquitin disassembly, and also binds linear and Lys-63 linked polyubiquitin with a lower affinity. Knock-down of USP5 causes the accumulation of p53/TP53 and an increase in p53/TP53 transcriptional activity because the uchored polyubiquitin that accumulates is able to compete with ubiquitinated p53/TP53 but not with MDM2 for proteasomal recognition.Specifications
USP5 | |
Monoclonal | |
Unconjugated | |
USP5 | |
ISOT | |
Mouse | |
Affinity Chromatography | |
RUO | |
297593, 486718, 8078 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P45974 | |
USP5 | |
Full length human recombit protein of human USP5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.