Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568358 Supplier Invitrogen™ Supplier No. PA568358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human, Mouse, and Rat samples.

USP51 (Ubiquitin Specific Peptidase 51) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include cysteine-type endopeptidase activity and thiol-dependent ubiquitinyl hydrolase activity. Specifically deubiquitinates 'Lys-14' (H2AK13Ub) and 'Lys-16'(H2AK15Ub) of histone H2A regulating the DNA damage response at double-strand breaks (DSBs). USP51 is recruited to chromatin after DNA damage and regulates the dynamic assembly/disassembly of TP53BP1 and BRCA1. Exhibits also activity for 'Lys-27' or 'Lys-63'-linked di-ubiquitin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP51
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP51
Gene Accession No. Q70EK9
Gene Alias AV136873; Deubiquitinating enzyme 51; RGD1562211; Ubiquitin carboxyl-terminal hydrolase 51; ubiquitin specific peptidase 51; ubiquitin specific protease 51; ubiquitin thioesterase 51; ubiquitin thiolesterase 51; ubiquitin-specific-processing protease 51; Usp51
Gene Symbols USP51
Host Species Rabbit
Immunogen Synthesized peptide derived from human USP51.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158880, 317398, 635253
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.