Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP51 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535274
Description
USP51 Polyclonal Antibody for Western Blot, ICC/IF
USP51 (Ubiquitin Specific Peptidase 51) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include cysteine-type endopeptidase activity and thiol-dependent ubiquitinyl hydrolase activity. Specifically deubiquitinates 'Lys-14' (H2AK13Ub) and 'Lys-16'(H2AK15Ub) of histone H2A regulating the DNA damage response at double-strand breaks (DSBs). USP51 is recruited to chromatin after DNA damage and regulates the dynamic assembly/disassembly of TP53BP1 and BRCA1. Exhibits also activity for 'Lys-27' or 'Lys-63'-linked di-ubiquitin.
Specifications
USP51 | |
Polyclonal | |
Unconjugated | |
USP51 | |
AV136873; Deubiquitinating enzyme 51; RGD1562211; Ubiquitin carboxyl-terminal hydrolase 51; ubiquitin specific peptidase 51; ubiquitin specific protease 51; ubiquitin thioesterase 51; ubiquitin thiolesterase 51; ubiquitin-specific-processing protease 51; Usp51 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
158880 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q70EK9 | |
USP51 | |
KLH conjugated synthetic peptide between 278-307 amino acids from the central region of human USP51. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction