Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP53 Antibody (OTI3C9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274812FR
Description
USP53 Monoclonal specifically detects USP53 in Human samples. It is validated for Western Blot.Specifications
USP53 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
USP53 | |
Recombinant protein fragment corresponding to amino acids 495-582 of human USP53 (NP_061923) produced in E.coli. | |
0.1 mL | |
54532.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3C9 | |
Western Blot | |
DKFZp781E1417, inactive ubiquitin carboxyl-terminal hydrolase 53, Inactive ubiquitin-specific peptidase 53, KIAA1350, ubiquitin specific peptidase 53, ubiquitin specific protease 53, ubiquitin specific proteinase 53 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction