Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP53 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557424 Supplier Invitrogen™ Supplier No. PA557424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APNGFKQHGN PHLYHSQGKG SYKHDRVVPQ SRASAQIISS SKSQILAPGE KITGKVKSDN GTGYDTDSSQ DSRDRGNSCD SSSKSRNR Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 66%.

Tight junction-associated protein that is involved in the survival of auditory hair cells and hearing. Maybe by modulating the barrier properties and mechanical stability of tight junctions. Has no peptidase activity (PubMed:14715245). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP53
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP53
Gene Accession No. Q70EK8
Gene Alias Inactive ubiquitin carboxyl-terminal hydrolase 53; inactive ubiquitin-specific peptidase 53; KIAA1350; ubiquitin specific peptidase 53; ubiquitin specific protease 53; ubiquitin specific proteinase 53; USP53
Gene Symbols USP53
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP53. Recombinant protein control fragment (Product #RP-95855).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54532
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.