Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP53 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | USP53 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
USP53 Polyclonal specifically detects USP53 in Human samples. It is validated for Western Blot.Specifications
USP53 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
54532 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
DKFZp781E1417, inactive ubiquitin carboxyl-terminal hydrolase 53, Inactive ubiquitin-specific peptidase 53, KIAA1350, ubiquitin specific peptidase 53, ubiquitin specific protease 53, ubiquitin specific proteinase 53 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 886-1073 of human USP53 (NP_061923.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title