Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP54 Antibody (OTI1G6), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274813MFV500
Description
USP54 Monoclonal antibody specifically detects USP54 in Human samples. It is validated for Flow CytometrySpecifications
USP54 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
159195 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1G6 | |
Flow Cytometry | |
bA137L10.3, bA137L10.4, C10orf29, USP54 ubiquitin specific peptidase 54 | |
Human recombinant protein fragment corresponding to amino acids 358-700 of human USP54 (NP_689799) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction