Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP54 Antibody (OTI1G6), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274813PEATT594
Description
USP54 Monoclonal specifically detects USP54 in Human samples. It is validated for Flow Cytometry.Specifications
USP54 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
USP54 | |
Recombinant protein fragment corresponding to amino acids 358-700 of human USP54 (NP_689799) produced in E.coli. | |
0.1 mL | |
159195.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1G6 | |
Flow Cytometry | |
bA137L10.3, bA137L10.4, C10orf29, USP54 ubiquitin specific peptidase 54 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction