Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103905 Supplier Invitrogen™ Supplier No. PA5103905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total USP54.

USP54 has no peptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP54
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP54
Gene Accession No. Q6IE24, Q70EL1, Q8BL06
Gene Alias 4930429G18Rik; AI115571; bA137L10.3; bA137L10.4; C030002J06Rik; C10orf29; Inactive ubiquitin carboxyl-terminal hydrolase 54; inactive ubiquitin-specific peptidase 54; ubiquitin specific peptidase 54; ubiquitin specific protease 54; ubiquitin specific proteinase 54; Usp54
Gene Symbols USP54
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP54(Accession Q70EL1), corresponding to amino acid residues E733-E783.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 159195, 408223, 78787
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.