Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP54 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA568215

Catalog No. PIPA568215


Only null left
Add to Cart

Description

Description

Predicted to react with Mouse and Rat samples.

USP54 has no peptidase activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP54
Polyclonal
Unconjugated
USP54
4930429G18Rik; AI115571; bA137L10.3; bA137L10.4; C030002J06Rik; C10orf29; Inactive ubiquitin carboxyl-terminal hydrolase 54; inactive ubiquitin-specific peptidase 54; ubiquitin specific peptidase 54; ubiquitin specific protease 54; ubiquitin specific proteinase 54; Usp54
Rabbit
Antigen affinity chromatography
RUO
159195, 408223, 78787
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6IE24, Q70EL1, Q8BL06
USP54
Synthesized peptide derived from human USP54.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.