Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP6 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP276350
Description
USP6 Polyclonal specifically detects USP6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
USP6 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
USP6 | |
Peptide with sequence C-HHIDLDVRTTLRNH corresponding to internal region (near N-Terminus) according to NP_004496.2. | |
0.1 mg | |
9098.0 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA Detection limit 1:4000 | |
Deubiquitinating enzyme 6, EC 3.1.2.15, EC 3.4.19.12, HRP1, hyperpolymorphic gene 1, Proto-oncogene TRE-2, TRE17, Tre2, Tre-2, tre-2 oncogene, TRE2USP6-short, ubiquitin carboxyl-terminal hydrolase 6, ubiquitin specific peptidase 6-, ubiquitin specific peptidase 6 (Tre-2 oncogene), ubiquitin specific protease 6 (Tre-2 oncogene), ubiquitin thioesterase 6, Ubiquitin thiolesterase 6, ubiquitin-specific protease USP6, Ubiquitin-specific-processing protease 6 | |
Goat | |
Antigen Affinity-purified | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction