Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP6 Monoclonal Antibody (3C3G2)
GREENER_CHOICE

Catalog No. FEMA550276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550276 Supplier Invitrogen™ Supplier No. MA550276
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP6 Monoclonal Antibody for ELISA, ICC/IF, Western Blot

USP6 is a deubiquitinase with an ATP-independent isopeptidase activity, cleaving at the C-terminus of the ubiquitin moiety. It catalyzes its own deubiquitination. In vitro, isoform2, but not isoform3, shows deubiquitinating activity. USP6 promotes plasma membrane localization of ARF6 and selectively regulates ARF6-dependent endocytic protein trafficking. It is able to initiate tumorigenesis by inducing the production of matrix metalloproteinases following NF-kappa-B activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP6
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3C3G2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene USP6
Gene Accession No. P35125
Gene Alias Deubiquitinating enzyme 6; HRP1; hyperpolymorphic gene 1; proto-oncogene TRE-2; TBC1D3 and USP32 fusion; TRE17; TRE2; Tre-2; TRESMCR; Ubiquitin carboxyl-terminal hydrolase 6; ubiquitin specific peptidase 6; ubiquitin specific peptidase 6 (Tre-2 oncogene); ubiquitin specific protease 6 (Tre-2 oncogene); ubiquitin thioesterase 6; ubiquitin thiolesterase 6; ubiquitin-specific protease USP6; ubiquitin-specific-processing protease 6; USP6; USP6-short
Gene Symbols USP6
Host Species Mouse
Immunogen Recombinant Human Ubiquitin carboxyl-terminal hydrolase 6 protein (348-535AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9098
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.