Learn More
Invitrogen™ USP6NL Polyclonal Antibody

Description
The antibody detects endogenous levels of total USP6NL protein.
Specifications
Specifications
| Antigen | USP6NL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | USP6NL |
| Gene Accession No. | Q92738 |
| Gene Alias | AI316785; Kiaa0019; mKIAA0019; novel TBC domain containing protein; related to the N-terminus of tre; RNTRE; RN-tre; TRE2NL; USP6 N-terminal like; USP6 N-terminal-like protein; Usp6nl; USP6NL-IT1 |
| Gene Symbols | USP6NL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.