Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP7 Antibody (03), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306427MFV610
Description
USP7 Monoclonal antibody specifically detects USP7 in Human samples. It is validated for Flow CytometrySpecifications
USP7 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
03 | |
Flow Cytometry | |
Deubiquitinating enzyme 7, EC 3.1.2.15, EC 3.4.19.12, HAUSPTEF1, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, ubiquitin carboxyl-terminal hydrolase 7, ubiquitin specific peptidase 7 (herpes virus-associated), ubiquitin specific protease 7 (herpes virus-associated), Ubiquitin thioesterase 7, Ubiquitin-specific-processing protease 7 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human USP7 (NP_003461.2; Lys208-Glu560). | |
0.1 mL | |
Apoptosis, Cell Biology, DNA Repair, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
7874 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction