Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP7 Antibody (BLR072G), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP278709
Description
USP7 Monoclonal specifically detects USP7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
USP7 | |
Monoclonal | |
Unconjugated | |
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA. with 0.09% Sodium Azide | |
Deubiquitinating enzyme 7, EC 3.1.2.15, EC 3.4.19.12, HAUSPTEF1, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, ubiquitin carboxyl-terminal hydrolase 7, ubiquitin specific peptidase 7 (herpes virus-associated), ubiquitin specific protease 7 (herpes virus-associated), Ubiquitin thioesterase 7, Ubiquitin-specific-processing protease 7 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Apoptosis, Cell Biology, DNA Repair, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
7874 | |
Store at 2-8°C/ 1 year from date of receipt | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
BLR072G | |
Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 20 μL/1 mg lysate | |
NP_003461.2, NP_003461.2 | |
USP7 | |
A peptide representing a region between residue 1 and 50. | |
100 μL | |
Primary | |
Human, Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction