Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP7 Monoclonal Antibody (OTI3C7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504064S


Description
USP7 Monoclonal antibody specifically detects USP7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
USP7 | |
Monoclonal | |
0.73 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q93009 | |
Mouse | |
Affinity chromatography | |
RUO | |
7874 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3C7 | |
Unconjugated | |
USP7 | |
2210010O09Rik; AA409944; AA617399; AU019296; AW548146; C80752; deubiquitinating enzyme 7; Hausp; Herpes virus-associated ubiquitin-specific protease; herpes-virus-associated ubiquitin-specific protease; herpesvirus-associated ubiquitin-specific protease; mHAUSP; protease; rHAUSP; TEF1; Ubiquitin carboxyl-terminal hydrolase 7; ubiquitin specific peptidase 7; ubiquitin specific peptidase 7 (herpes virus-associated); ubiquitin specific protease 7; ubiquitin specific protease 7 (herpes virus-associated); ubiquitin thioesterase 7; ubiquitin thiolesterase 7; ubiquitin-specific-processing protease 7; UBP; USP7 | |
Full length human recombinant protein of human USP7 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction