Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589302 Supplier Invitrogen™ Supplier No. PA589302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSKKHTGYVG LKNQGATCYM NSLLQTLFFT NQLRKAVYMM PTEGDDSSKS VPLALQRVFY ELQHSDKPVG TKKLTKSFGW ETLDSFMQHD VQELCRVLLD NVENKMKGTC VEGTIPKLFR GKMVSYIQCK EVDYRSDRRE DYYDIQLSIK GKKNIFESFV DYVAVEQLDG DNKYDAGEHG LQEAEKGVKF LTLPPVLHLQ LMRFMYDPQT DQNIKINDRF EFPEQLPLDE FLQKTDPKDP ANYILHAVLV HSGDNHGGHY VVYLNPKGDG KWCKFDDDVV SRCTKEEAIE HNYGGHDDDL SVRHCTNAYM LVYIRESKLS EVLQAVTDHD IPQQLVERLQ EEKRIEAQKR KERQEAHLYM QVQIVAEDQF CGHQGNDMYD EEKVKYTVFK VLKNSSLAEF VQSLSQTMGF PQDQIRLWPM QARSNGTKRP AMLDNEADGN KTMIELSDNE NPWTIFLETV DPELAASGAT LPKFDKDHDV MLFLKMYDPK TRSLNYCGHI YTPISCKIRD LLPVMCDRAG FIQDTSLILY EEVKPNLTER IQDYDVSLDK ALDELMDGDI IVFQKDDPEN DNSELPTAKE YFRDLYHRVD VIFCDKTIPN DPGFVVTLSN RMNYFQVAKT VAQRLNTDPM LLQFFKSQGY RDGPGNPLRH NYEGTLRDLL QFFKPRQPKK LYYQQLKMKI TDFENRRSFK CIWLNSQFRE EEITLYPDKH GCVRDLLEEC KKAVELGEKA SGKLRLLEIV SYKIIGVHQE DELLECLSPA TSRTFRIEEI PLDQVDIDKE NEMLVTVAHF HKEVFGTFGI PFLLRIHQGE HFREVMKRIQ SLLDIQEKEF EKFKFAIVMM GRHQYINEDE YEVNLKDFEP QPGNMSHPRP WLGLDHFNKA PKRSRYTYLE KAIKIHN; Positive Samples: HepG2, Rat testis; Cellular Location: Cytoplasm, Nucleus, PML body.

USP7 or HAUSP is a ubiquitin specific protease or a deubiquitylating enzyme that cleaves ubiquitin from its substrates. Since ubiquitylation (polyubiquitination) is most commonly associated with the stability and degradation of cellular proteins, HAUSP activity generally stabilizes its substrate proteins. HAUSP is most popularly known as a direct antagonist of Mdm2, the E3 ubiquitin ligase for the tumor suppressor protein, p53. Normally, p53 levels are kept low in part due to Mdm2-mediated ubiquitylation and degradation of p53. Interestingly, in response to oncogenic insults, HAUSP can deubiquitinate p53 and protect p53 from Mdm2-mediated degradation, indicating that it may possess a tumor suppressor function for the immediate stabilization of p53 in response to stress. Another important role of HAUSP function involves the oncogenic stabilization of p53. Oncogenes such as Myc and E1A are thought to activate p53 through a p19 alternative reading frame (p19ARF, also called ARF)-dependent pathway, although some evidence suggests ARF is not essential in this process. An intriguing possibility is that HAUSP provides an alternative pathway for safeguarding the cell against oncogenic insults.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene USP7
Gene Accession No. Q4VSI4, Q6A4J8, Q93009
Gene Alias 2210010O09Rik; AA409944; AA617399; AU019296; AW548146; C80752; deubiquitinating enzyme 7; Hausp; Herpes virus-associated ubiquitin-specific protease; herpes-virus-associated ubiquitin-specific protease; herpesvirus-associated ubiquitin-specific protease; mHAUSP; protease; rHAUSP; TEF1; Ubiquitin carboxyl-terminal hydrolase 7; ubiquitin specific peptidase 7; ubiquitin specific peptidase 7 (herpes virus-associated); ubiquitin specific protease 7; ubiquitin specific protease 7 (herpes virus-associated); ubiquitin thioesterase 7; ubiquitin thiolesterase 7; ubiquitin-specific-processing protease 7; UBP; USP7
Gene Symbols USP7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 206-1102 of human USP7/HAUSP (NP_0034612).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252870, 360471, 7874
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.