Learn More
USP8 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302929AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
USP8 is a ubiquitin specific protease that plays an important regulatory role at the level of protein turnover by preventing degradation. USP8 is involved in cell proliferation, and probably regulates the stability of STAM2 and RASGRF1. USP8 may regulate T-cell anergy mediated by RNF128 via the formation of a complex containing RNF128 and STAM2. As revealed by structur/function studies, USP8 forms a ternary complex with RNF128 and OTUB1, and interacts with the SH3 domain of STAM2 and RASGRF1. Expression of USP8 is induced Upon growth stimulation in starved human fibroblasts, and expression decreases in response to growth arrest induced by cell-cell contact.Specifications
USP8 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P40818 | |
USP8 | |
Between 400 and 450 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
USP8 | |
deubiquitinating enzyme 8, hUBPy, HumORF8, KIAA0055, SPG59, ubiquitin isopeptidase Y, ubiquitin specific protease 8, Ubiquitin thioesterase 8, ubiquitin thiolesterase 8, ubiquitin-specific-processing protease 8, UBPY | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9101 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.