Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP9x Antibody (OTI2G7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274822B
Description
USP9x Monoclonal specifically detects USP9x in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
USP9x | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
USP9X | |
Recombinant protein fragment corresponding to amino acids 2246-2570 of human USP9x (NP_001034680) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2G7 | |
Western Blot, Immunohistochemistry | |
Deubiquitinating enzyme FAF-X, DFFRXubiquitin specific protease 9, X-linked (fat facets-like, Drosophila), Drosophila fat facets related, X-linked, EC 3.1.2.15, EC 3.4.19.12, FAF, FAM, Fat facets in mammals, fat facets protein related, X-linked, Fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific peptidase 9, X-linked, ubiquitin specific peptidase 9, X-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-X, Ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, Ubiquitin-specific protease 9, X chromosome, Ubiquitin-specific-processing protease FAF-X, USP9, X chromosome (fat facets-like Drosophila) | |
Mouse | |
Affinity Purified | |
Apoptosis, Autophagy, Cancer, Neuroscience | |
8239.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction