Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP9X Monoclonal Antibody (4B7)
GREENER_CHOICE

Catalog No. PIMA520360
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520360 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520360 Supplier Invitrogen™ Supplier No. MA520360
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MTATTRGSPV GGNDNQGQAP DGQSQPPLQQ NQTSSPDSSN ENSPATPPDE QGQGDAPPQL EDEEPAFPHT DLAKLDDMIN RPRWVVPVLP.

USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP9X
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4B7
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene USP9X
Gene Accession No. Q93008
Gene Alias 5730589N07Rik; AA407302; AA407699; AL022658; AL022749; Deubiquitinating enzyme FAF-X; DFFRX; Drosophila fat facets related, X-linked; FAF; Fafl; FAF-X; FAM; fat facets homolog; Fat facets in mammals; fat facets protein related, X-linked; fat facets protein-related, X-linked; fats facets protein related, X; hFAM; MRX99; MRXS99F; Probable ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-x-like protein; RGD1560056; Ubiquitin carboxyl-terminal hydrolase FAM; ubiquitin specific peptidase 9 X-linked; ubiquitin specific peptidase 9, X chromosome; ubiquitin specific peptidase 9, X-linked; ubiquitin specific protease 9 SSSRF- isoform; ubiquitin specific protease 9, X chromosome; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); ubiquitin specific protease 9, X-linked; ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X; ubiquitin-specific protease 9, X chromosome; Ubiquitin-specific-processing protease FAF-X; USP9; USP9X
Gene Symbols USP9X
Host Species Mouse
Immunogen USP9X (NP_068706, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8239
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.